missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Solution, APHA for Metals, Ricca Chemical

Catalog No. 428516
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
428516 500 mL Natural Poly Bottle
428532 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 428516 Supplier Ricca Chemical Company Supplier No. 428516
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0 , 1312-81-8

Specifications

Packaging Natural Poly Bottle
CAS Min % 72.0, 21.35, 4.27
CAS Max % 75.0, 22.22, 4.45
Methods APHA for Metals
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Quantity 500 mL
Chemical Name or Material Lanthanum Solution