missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 50 mL |
| Concentration or Composition (by Analyte or Components) | Nitric acid: 5.0%; Lanthanum(III) oxide: 0.12%; Water: 94.88% |
| Chemical Name or Material | Lanthanum |
Safety and Handling
| Hazard Category | H290-H314-H335 |
| Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
| DOTInformation | Hazard Class: 8, Packing Group: III |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only