missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Nitrate TS, Ricca Chemical

Catalog No. R4289500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4289500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4289500 Supplier Ricca Chemical Company Supplier No. R4289500500A

Chemical Identifiers

CAS 10277-43-7 , 7732-18-5
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
PubChem CID 16211478
IUPAC Name lanthanum(3+) hexahydrate trinitrate
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 4 to 5%
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Infinitely soluble in water
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 500 mL
Percent Purity 0.05
Grade USP
Chemical Name or Material Lanthanum Nitrate TS, Hexahydrate
Show More Show Less