missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum ICP Standard, 1000 ppm La in 3% HNO3, Ricca Chemical

Catalog No. p-3251848
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PLA1KN100 Natural LDPE Bottle
PLA1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PLA1KN100 Supplier Ricca Chemical Company Supplier No. PLA1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1312-81-8 , 7697-37-2 , 7732-18-5

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm La
CAS Min % 93.0, 4.62, 0.11
CAS Max % 97.0, 4.81, 0.11
Equivalence 1 mL = 1 mg La
Chemical Name or Material Lanthanum ICP Standard