missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019367
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019367 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019367 Supplier Merck Emd Millipore Supplier No. PHR13921G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lamotrigine is an anticonvulsant drug, which is widely used in the treatment of epilepsy and bipolar disorders. It is also used in combination with other antiepileptics to treat refractory partial seizures, generalized tonic-clonic seizures and generalized toxic-chronic seizures.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 84057-84-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C9H7 Cl2N5
MDL Number MFCD00865333
Synonym Lamotrigine; 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine; GI 267119 X
Type Certified Reference Material
Quantity 1 g
Formula Weight 256.09
Grade Certified Reference Material