missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine, 98%

Catalog No. AC460850050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460850050 5 g
AC460850010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460850050 Supplier Thermo Scientific Chemicals Supplier No. 460850050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84057-84-1 | C9H7Cl2N5 | 256.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84057-84-1
Molecular Formula C9H7Cl2N5
Molecular Weight (g/mol) 256.09
MDL Number MFCD00865333
InChI Key PYZRQGJRPPTADH-UHFFFAOYSA-N
Synonym lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
PubChem CID 3878
ChEBI CHEBI:6367
IUPAC Name 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
SMILES NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 218.0°C
Loss on Drying 0.5% max. (105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmf
Formula Weight 256.09
Percent Purity 98%
Chemical Name or Material Lamotrigine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 281-901-8

RUO – Research Use Only