missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Laminin Mouse Protein, Natural

Numéro de catalogue. 23017015
Encompass
Modifier l'affichage
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
23017015 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 23017015 Fournisseur Gibco™ Code fournisseur 23017015
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Laminin Mouse Protein, Natural is the major glycoprotein component of basement membranes, and has a molecular weight of 850,000 daltons.

Laminin is the major glycoprotein component of basement membranes, and has a molecular weight of 850,000 daltons. This preparation of mouse Laminin was isolated from the Engelbreth-Holm-Swarm (EHS) sarcoma and is supplied at a concentration of 0.5-2.0 mg/mL.

Key functions include:

  • Modulation of cell attachment, cell spreading, cell growth and motility
  • Binding to itself and other matrix compounds
  • Promotion of epithelial differentiation
  • Modification of leukocyte function
  • Stimulation of neurite outgrowth

Supplied: 1 mg in 50 mM Tris-HCL (pH 7.4), 0.15 M NaCl

Performance qualification (cell attachment): Half-maximal cell attachment of MG63 cells at 37°C: ≤20 μg/mL

Spécifications

À utiliser avec (application) Cell Culture
Quantité 1 mg
Emballage Vial
Alias du gène Laminin
Type de produit Mouse laminin
Conjugué Unconjugated
Espèces Mouse
Recombinant Native
Stérilité Sterile
Contenu et stockage Store in ultra-cold freezer (-68 to -85°C) and protect from light.
Shipping condition (Condition d'expédition) Dry Ice
Forme Frozen
Sous-type de protéine Laminin
Catégorie de recherche Extracellular Matrix & Cell Adhesion, Stem Cell Research
Couleur Clear
Afficher plus de résultats Afficher moins de résultats

For Research Use Only. Not for use in diagnostic procedures.