missing translation for 'onlineSavingsMsg'
Learn More

Lactitol monohydrate, 98%

Catalog No. AAB2101606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2101606 5 g
AAB2101603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2101606 Supplier Thermo Scientific Chemicals Supplier No. B2101606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 81025-04-9 | C12H26O12 | 362.328 g/mol

Lactitol monohydrate is used in low fat foods, anthelmintic, cosmetic and pharmaceutical excipients. It is used in sugar-free candies, cookies, chocolate and ice cream. It is a laxative, which used to treat constipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lactitol monohydrate is used in low fat foods, anthelmintic, cosmetic and pharmaceutical excipients. It is used in sugar-free candies, cookies, chocolate and ice cream. It is a laxative, which used to treat constipation.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 81025-04-9
Molecular Formula C12H26O12
Molecular Weight (g/mol) 362.328
MDL Number MFCD00150767
InChI Key LXMBXZRLTPSWCR-XBLONOLSSA-N
Synonym lactitol monohydrate, d-lactitol monohydrate, lactitol hydrate, unii-uh2k6w1y64, 4-o-beta-d-galactopyranosyl-d-glucitol monohydrate, d-glucitol, 4-o-beta-d-galactopyranosyl-, monohydrate, 4-o-beta-d-galactopyranosyl-d-glucitol, portolac tn, 2s,3r,4r,5r-4-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyhexane-1,2,3,5,6-pentol;hydrate
PubChem CID 3067270
IUPAC Name (2S,3R,4R,5R)-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol;hydrate
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O.O

Specifications

Melting Point 95°C to 98°C
Quantity 5 g
Merck Index 14,5340
Solubility Information Soluble in water.
Formula Weight 362.33 (344.31 Anhydrous)
Percent Purity 98%
Chemical Name or Material Lactitol monohydrate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only