missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactitol, ≥99% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 585-86-4 |
| Linear Formula | C12H24O11 |
| MDL Number | MFCD00079407 |
| Synonym | 4-O-beta-D-Galactopyranosyl-D-glucitol; D-Lactitol; E 966; Lactobiosit; Lactositol; no translation |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 344.31 |
| Percent Purity | ≥99% (HPLC) |
| Grade | Analytical Standard |