missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide lactique, certifié, 1,00N ±0,05N (1,0M), LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Lactic Acid |
| Couleur | Incolore |
| CAS % maximal | 91.15, 8.85 |
| Formule linéaire | C3H6O3 |
| Renseignements sur la solubilité | Soluble in water |
| Concentration | 1.00N ±0.05N (1.0M) |
| Poids de formule | 90.08 |
| Niveau | Certifié |
| Densité | 1.02g/mL |
| Quantité | 500 mL |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Température ambiante |