missing translation for 'onlineSavingsMsg'
Learn More

Acide lactique, grade ACS, 85,0 à 90,0%, LabChem™

Numéro de catalogue. LC159501
Modifier l'affichage
Click to view available options
Quantité:
500 mL
Emballage:
Verre ambré
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
LC159501 500 mL Verre ambré
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. LC159501 Fournisseur LabChem Code fournisseur LC159501

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 50-21-5 , 7732-18-5
Formule moléculaire C3H6O3
Poids moléculaire (g/mol) 90.08
Numéro MDL MFCD00004520
Clé InChI JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonyme lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
Nom de l’IUPAC Acide 2-hydroxypropanoïque
SOURIRES CC(O)C(O)=O

Spécifications

Nom ou substance chimique Lactic Acid
Point de fusion 16,8 °C
Point d’ébullition 122 °C
Couleur Incolore
CAS % maximal 85, 15
Quantité 500 mL
Point d’éclair 113 °C
Emballage Verre ambré
Forme physique Liquide
Formule linéaire C3H6O3
Numéro UN UN3265
Renseignements sur la solubilité Soluble dans l’eau
Poids de formule 90.08
Pourcentage de pureté 85,0 à 90,0%
Niveau ACS
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep only in original container.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 200-018-0
Recommended Storage Température ambiante