Learn More
Lactic acid, ACS, 85.0-90.0% aq. soln.
Description
It is used most commonly by athletes to describe the intense pain felt during exhaustive exercise. Used to make cultured dairy products, as a food preservative, and to make chemicals. Useful for the creation of highly aligned electrospun poly-L-lactic acid fibers for nerve regeneration applications. Poly(lactic acid) nanocomposites with various organoclays shows useful application for gas permeability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Melting Point | 18°C |
| Boiling Point | 122°C (15 mm) |
| Density | 1.209 g/mL |
| Quantity | 100 g |
| Flash Point | >110°C (230°F) |
| Physical Form | Liquid |
| Refractive Index | 1.427 |
| Assay Percent Range | 85.0 to 90.0% |
| Beilstein | 1209341 |
| Merck Index | 14,5336 |
| Show More |
Safety and Handling
| Hazard Category | H315-H318 |
| Hazard Statement | GHS H Statement H318-H315 |
| Precautionary Statement | P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390 |
| EINECSNumber | 200-018-0 |
| RTECSNumber | OD2800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only