missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 20% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 42722.5

Quantity 10 L
Packaging Cubitainer

 View more versions of this product

Catalog No. 427225

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

50-21-5 , 7732-18-5
90.08
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
612
2-hydroxypropanoic acid
C3H6O3
MFCD00004520
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
CHEBI:78320
CC(O)C(O)=O

Specifications

Lactic Acid
80.0, 18.2
C3H6O3
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
CC(O)C(O)=O
90.08
CHEBI:78320
Cubitainer
50-21-5 , 7732-18-5
83.0, 18.94
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
2-hydroxypropanoic acid
612
10 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents