missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 1.00 N, Ricca Chemical

Catalog No. 428032
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
428032 1 L Natural Poly Bottle
428016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 428032 Supplier Ricca Chemical Company Supplier No. 428032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) Mixture
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

CAS Min % 89.0, 9.12
Color Colorless
CAS Max % 93.0, 9.49
Concentration 1.00 Normal
Quantity 1 L
Packaging Natural Poly Bottle
Physical Form Liquid
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
Grade Laboratory
Chemical Name or Material Lactic Acid