missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 1.00 N, Ricca Chemical

Catalog No. 428016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
428016 500 mL Natural Poly Bottle
428032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 428016 Supplier Ricca Chemical Company Supplier No. 428016

Chemical Identifiers

CAS 7732-18-5 , 50-21-5

Specifications

Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1.00N
Quantity 500 mL
Packaging Natural Poly Bottle
Chemical Name or Material Lactic Acid