missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ LabX™ Melting Point Express Starter Pack

Catalog No. 01915036
Change view
Click to view available options
For Use With (Equipment):
Melting Point Excellence Systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01915036 Melting Point Excellence Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01915036 Supplier Mettler Toledo™ Supplier No. 30005779

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Connect up to three Mettler Toledo™ instruments at once to seamlessly perform complex workflows in melting, boiling, cloud, and slip melting point. Includes one Thermal Values instrument license.

  • Seamlessly perform complex workflows in melting, boiling, cloud, and slip melting point
  • Software automatically stores results and enables users to effectively manage large amounts of data
  • LabX Thermal Values allows users to workflows to match standard operating procedures
  • Complies with FDA 21 CFR Part 11 regulations to be audit-ready at anytime with a secure, and traceable database
  • Convenient search filters and sorting facilitate the retrieval of previous measurements
  • Includes one Thermal Values instrument license
  • Intensity curves and videos are available
  • Reports can be personalized and printed

Specifications

Type LabX™ Starter Pack (Express Edition)
Display Type No Display
Description Full version of LabX for installation on a single computer
Includes 1 Thermal Values Instrument License
For Use With (Equipment) Melting Point Excellence Systems