missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 500 mL with 2 Ports, Luer Lock, 2D BPC
GREENER_CHOICE

Simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Supplier:  Thermo Scientific™ SH30657.14

 Afficher plus de versions de ce produit

Numéro de catalogue. SH3065714


Il en reste null

Description

Description

Using Thermo Scientific™ Labtainer™ BPC bag with 2 top ports (luer lock and MPC insert), simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance, and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation systems

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport &

    Shipping - bulk sterile liquids, buffers, media and process intermediates within and between facilities

Spécifications

Identifiants chimiques

500 mL
Packages placed in strong cardboard cartons with product and lot identification
Gamma Irradiated
Edge Ports: 2 x 0.25 in.
26.4 cm x 17.3 cm (10.4 in. x 6.8 in.)
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Labtainer
CX5-14

Spécifications

500 mL
Packages placed in strong cardboard cartons with product and lot identification
10 per case
Line 1. Body luer lock connection, polypropylene. No Tubing
Labtainer
CX5-14
26.4 cm x 17.3 cm (10.4 in. x 6.8 in.)
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Gamma Irradiated
Line 2. Insert luer lock connection, polypropylene. No Tubing Line
Edge Ports: 2 x 0.25 in.
No Tubing
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions