missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors with clamps, injection site
GREENER_CHOICE

Catalog No. SS00017I
Change view
Click to view available options
Quantity:
25/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SS00017I 25/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SS00017I Supplier Thermo Scientific™ Supplier No. SS00017I

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

Specifications

Content And Storage Store at room temperature.
Includes 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Material EVA
Sterility Sterile
Tubing Style EVA
Film Description ASI™ 28
Outer Packaging One 4-mil. poly bag
Port Detail MPC insert and body connectors, plug and injection site
Product Line Labtainer
Quantity 25/Cs.
Shipping Condition Room Temperature
Sterilization Method Irradiated
Dimensions (L x W x H) 56.6 x 32.5 cm (22.3 x 12.8 in.)
Width (English) 12.8 in.
Product Type Bioprocess Container
Capacity (Metric) 10 L
Length (Metric) 56.6 cm
Width (Metric) 32.5 cm
Show More Show Less