Learn More
MilliporeSigma™ Labscale TFF System Reservoir
Numéro de catalogue. p-117415
Change view
Click to view available options
Type:
Reservoir
Numéro de catalogue. Type
XX42RES01 Reservoir
1 options
Numéro de catalogue. XX42RES01 Fournisseur MilliporeSigma™ Code fournisseur XX42RES01
Il en reste null
Comprend
Graduated 500mL acrylic reservoir with retentate back pressure valve, feed and retentate pressure indicators; stirrer assembly; diaphragm pump assembly with tubing and power supply.
Easy to use for sample preparation
Complete linear, scalable solution for small-volume processing designed specifically to use the Pellicon™ XL Device for simple sample prep and more complex scale-up work.
  • 500mL reservoir accepts direct docking of the Pellicon XL device, eliminating need for tubing connections
Order Info
Individual components are also available for a custom small-volume TFF system. Pellicon™ XL devices sold separately.
Spécifications
For Use With (Equipment) Labscale TFF System
Material Acrylic
For Use With (Application) Accepts direct docking of the Pellicon™ XL device, eliminating need for tubing connections
Includes Retentate back pressure valve
Quantity 1/Pk.
Type Reservoir
Volume (Metric) 500 mL