missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ VACUUBRAND™ Hybrid™ Vacuum Pump Oil

Catalog No. 10030181
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10030181 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10030181 Supplier Labconco™ Supplier No. 7584300

missing translation for 'validMainframeMsg'

For VACUUBRAND pumps

VACUUBRAND™ B-Oil. One. liter bottle. Replacement oil for VACUUBRAND HYBRID™ Vacuum Pumps 7584000 and 7584002.

Specifications

Product Type Oil
Format Oil
For Use With (Equipment) Hybrid Vacuum Pumps
Quantity 1 L
Volume (English) 33.8 oz.
Volume (Metric) 1 L