Learn More
Labconco™ Transition Connectors for Downdraft Powder Stations
Description
Epoxy-coated steel bracket and fasteners connect two Protector Downdraft Powder Stations together, extending the width of the work surface area and creating a continuous work surface.
Specifications
Specifications
| For Use With (Equipment) | Protector Downdraft Powder Station |
| Weight (English) | 15 lb. |
| Weight (Metric) | 6.8 Kg |
| Material | Epoxy-coated Steel |
| For Use With (Application) | Latent Fingerprint Development |