Learn More
Labconco™ Syringe Tubes
Description
Accessory to FilterMate Portable Exhausters and XPert Balance Systems.
- Formaldehyde and ammonia contaminant detector tubes indicate the presence of a specific set of contaminants with a color change
- Detector tubes for organic vapors may be ordered from a laboratory supply distributor
Spécifications
Spécifications
| For Use With (Equipment) | FilterMate Portable Exhauster (2005 and Newer), XPert Balance System |
| Weight (English) | 1 lb. |
| Weight (Metric) | 0.5 Kg |
| Product Type | Ammonia Detector Tubes |
| For Use With (Application) | Carbon Saturation Detection |