Learn More
Labconco™ Syringe Tubes
Catalog No. 1030485
Change view
Click to view available options
Product Type:
Ammonia Detector Tubes
Formaldehyde Detector Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
1030485 Ammonia Detector Tubes
1030484 Formaldehyde Detector Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1030485 Supplier Labconco™ Supplier No. 6925700
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For carbon saturation detection
Accessory to FilterMate Portable Exhausters and XPert Balance Systems.
  • Formaldehyde and ammonia contaminant detector tubes indicate the presence of a specific set of contaminants with a color change
  • Detector tubes for organic vapors may be ordered from a laboratory supply distributor
Specifications
For Use With (Equipment) FilterMate Portable Exhauster (2005 and Newer), XPert Balance System
Weight (English) 1 lb.
Weight (Metric) 0.5 Kg
Product Type Ammonia Detector Tubes
For Use With (Application) Carbon Saturation Detection