Learn More
Labconco™ Support Shelves: Telescoping Base Stand
For use with Telescoping Base Stands supporting Purifier Logic Biosafety Cabinets and PuriCare Procedure Stations.
Supplier:  Labconco™ 3811101
 Afficher plus de versions de ce produit
Numéro de catalogue. 10269304
Il en reste null
Description
Description
For use with Telescoping Base Stands supporting Purifier Logic Biosafety Cabinets and PuriCare Procedure Stations (pre-2014). Support Shelf provides 8.0 in. x 20.0 in. (20 x 51 cm) work surface to support remote equipment such as an aspirator pump within easy access of the biosafety cabinet. Powder-coated steel shelf mounts to left-side tubular frame of the base stand. Includes powder-coated steel shelf and fasteners. Not for use with SoLo Hydraulic Lift Base Stand. Installation is required.
Spécifications
Identifiants chimiques
Support Shelf
Spécifications
3 ft. Telescoping Base Stand
5 Kg
36 in.
91.44 cm
Powder-coated Steel
12 lb.
9.9 in.
25.14 cm
Support Shelf
Provides Work Surface to Support Remote Equipment
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS