Learn More
Labconco™ Rotors for CentriVap™ Systems
For use with Labconco CentriVap Systems
Supplier: Labconco™ 7461900
Description
For use with Labconco CentriVap Systems
- Anodized aluminum rotors hold tubes at angle to maximize surface area
- Two hole sizes on rotors 16-317-54 and 16-317-62 to -64 accommodate tubes with different O.D.
- Maximum tube capacity for a given O.D. may vary with overall tube height
- CentriVap DNA Systems and Proteomic come with DNA Rotor (16-317-54); Acid Systems come with Acid-Resistant 12-13 mm Rotor (16-108-450)
To prevent loss of sample during centrifugation, fill tubes only 0.33 to 0.5 full.
Identifiants chimiques
| Anodized Aluminum | |
| CentriVap™ DNA system |
Spécifications
| Anodized Aluminum | |
| CentriVap™ DNA system | |
| 3 lb. |
| Microtiter Plate Rotor | |
| 4 Standard or 2 Deep-well Plates | |
| 1.4 kg |