Learn More
Labconco™ Replacement Lid Gasket
Replacement lids for use with FreeZone 1, 2.5 & 4.5 Liter Freeze Dry Systems.
Supplier:  Labconco™ 7688500
Numéro de catalogue. LCN7688500
Il en reste null
Description
Description
Replacement lids for use with FreeZone 1, 2.5 & 4.5 Liter Freeze Dry Systems.
Replacement rubber gasket allows the chamber lid of a FreeZone Freeze Dryer to seal to the chamber during the lyophilization process.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS