missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Nonventilated PCR and Tissue Culture Enclosures

Catalog No. 16108286
Change view
Click to view available options
Lighting Type:
Fluorescent, UV
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lighting Type
16108286 Fluorescent, UV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Because these enclosures do not provide protection to the user, they should not be used in conjunction with biohazardous material, toxins or radionuclides. You and your safety officer must carefully assess the risk associated with any operation performed in a Purifier™ nonventilated enclosure.
Catalog No. 16108286 Supplier Labconco™ Supplier No. 3952400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Because these enclosures do not provide protection to the user, they should not be used in conjunction with biohazardous material, toxins or radionuclides. You and your safety officer must carefully assess the risk associated with any operation performed in a Purifier™ nonventilated enclosure.

Includes

PCR enclosures include variable timer for UV exposure with eight settings in minutes of 5, 10, 15, 30, 60, 120, 240 and continuous.

Requires

Supporting base, work surface (available separately).

Designed to reduce cross-contamination during PCR procedures and culture manipulations

  • UV-resistant, glacier white and gray epoxy-coated aluminum and steel construction
  • UV-opaque and resistant, 0.25 in. (0.6cm) thick tempered safety glass sash and sides
  • Ergonomic 20° angled and hinged sash that pivots upward, locks to a loading height of 20 in. (50.8cm) and has wiping seals to reduce cross-contamination
  • 7.8 in. (19.8cm) High sash opening
  • UV lamp w/digital timer and interlock UV protection panel
  • Fluorescent lamp
  • 254nm UV lamp
  • Interior electrical receptacle
  • Front-mounted switches for three-way light (fluorescent, UV and off) and auxiliary electrical receptacle
  • Two utility ports with iris openings and plugs

Order Info

48W x 26.75D x 25.75 in.H (121.9 x 190.5 x 65.4cm); Shipping weight: 100 lb. (45kg); Airflow monitor not included

Certifications

ETL listed (115V models); CE marked (230V model)

Compliance

UL 61010A-1, CAN/CSA C22.2 No. 1010.1, CE

Warranty

One year

Specifications

Product Type PCR Enclosure
Coating Epoxy
Lighting Type Fluorescent, UV
Material Epoxy-coated Aluminum and Steel
Amperage 10 A
Certifications/Compliance UL, CAN/CSA, ETL, CE
Depth (English) 26.7 in.
Depth (Metric) 67.8 cm
For Use With (Application) Designed to reduce cross-contamination during PCR procedures and culture manipulations
Height (English) 25.7 in.
Height (Metric) 65.2 cm
Height (Metric) Sash Opening 19.8 cm
Voltage 115 V
Weight (English) 100 lb.
Weight (Metric) 45 kg
Width (English) 48 in.
Width (Metric) 121.9 cm
Frequency 60 Hz
Show More Show Less