Learn More
Labconco™ Protector™ Downdraft Powder Station
The Labconco Protector™ Downdraft Powder Station keeps users safe by containing powders during fingerprint processing.
$7,855.71 - $9,415.36
Spécifications
| Product Type | Downdraft Powder Station |
|---|---|
| Coating | Epoxy Powder |
| Filter Type | HEPA |
| Material | Stainless Steel |
| Depth (English) | 25.7 in. |
Avertissement:
Does not provide protection from biohazardous aerosols or gaseous contaminants. Contact your Fisher Customer Service Representative for information on biosafety cabinets and fume hoods if your application involves these contaminants.
| Numéro de catalogue | Numéro du manufacturier. | Voltage | Width (Metric) | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Voltage | Width (Metric) | Prix | Quantity & Availability | ||||
|
09681301
|
Labconco™
3410000 |
115 V | 60.9 cm |
chaque for $7,855.71
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
09681305
|
Labconco™
3410001 |
115 V | 91.4 cm |
chaque for $9,415.36
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Open-sided, open-top workstation provides unrestricted operator movement and accommodates oversized evidence.
- Powder-coated steel housing and shelf
- Type 304 stainless steel work surface with Flow-Zone™ perforations
- Built-in blower with motorized, backward curved fan and switch
- With removable powder catch pan
- Exhaust prefilter of pleated paper with MERV 8 rating
- Exhaust HEPA filter, 99.99% efficient on particles 0.3 micron
- Red, filter system alarm light alerts operator to low airflow conditions
- Comes with 6 ft. (15cm) power cord with plug
- Available in 2 ft. (0.6m) and 3 ft. (0.9m) widths and Domestic electrical standards
An optional Transition Connector mounts between two downdraft powder hoods for a secure fit and continuous work surface.
Compliance
ETL listed (Models 3410000 and 3410001).
Spécifications
| Downdraft Powder Station | |
| HEPA | |
| 25.7 in. | |
| 18.75 in. |
| Epoxy Powder | |
| Stainless Steel | |
| 65.3 cm | |
| 47.6 cm |