Learn More
Labconco™ Protector™ Acid and Standard Storage Cabinets Shelf Kits
Catalog No. 1630545
Change view
Click to view available options
For Use With (Equipment):
Protector Acid Storage Cabinet
Protector Standard Storage Cabinet
Width (English) Exterior:
24 in.
30 in.
36 in.
48 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Width (English) Exterior
1630545 Protector Acid Storage Cabinet 30 in.
1630540 Protector Standard Storage Cabinet 48 in.
1630542 Protector Acid Storage Cabinet 24 in.
1630543 Protector Acid Storage Cabinet 36 in.
1630544 Protector Acid Storage Cabinet 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 1630545 Supplier Labconco™ Supplier No. 9919002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Shelf Kits provide additional storage space in Labconco Protector Standard and Acid Base Cabinets.
Interior Shelf Kit provides additional storage space inside Labconco Protector Acid Storage Cabinets. Includes powder-coated steel shelf, acid-resistant PVC tray, and mounting hardware.
Specifications
For Use With (Equipment) Protector Acid Storage Cabinet
Width (Metric) Exterior 76 cm
Product Type Shelf Kit
Includes Epoxy Coated Steel shelf, Acid-resistant Polyethylene tray and mounting hardware
Width (English) Exterior 30 in.