Learn More
Labconco™ Manual Valve Gas and Vacuum Tubing Connection Kit for Precise™ Glove Boxes
Description
- Includes flexible vinyl tubing and fittings
- Connects user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Specifications
Specifications
| For Use With (Equipment) | Precise Basic Glove Box, Controlled Atmosphere Glove Box |
| Weight (English) | 20 lb. |
| Weight (Metric) | 9.1 Kg |
| Product Type | Manual Valve Gas and Vacuum Tubing Connection Kit |