Learn More
Labconco™ Class II Cabinets IV Bar Kit
Bar supports intravenous solution bottles and bags in Purifier Biosafety Cabinets.
Supplier:  Labconco™ 3858602
 Afficher plus de versions de ce produit
Numéro de catalogue. 10269252
Il en reste null
Description
Description
Bar supports intravenous solution bottles and bags. Kit includes IV bar, mounting hardware and four hangers. For use with Labconco Purifier Class II Biosafety Cabinets. Installation is required.
Spécifications
Identifiants chimiques
Exhaust Safety Cabinets Kit
Spécifications
Exhaust Safety Cabinets Kit
6 lb.
2.72 kg
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS