missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Poly Drum with Screw-on Lid
GREENER_CHOICE

Catalog No. 19338482
Change view
Click to view available options
Capacity (English):
20 gal.
30 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19338482 20 gal.
19338484 30 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19338482 Supplier Eagle™ Supplier No. 1650BEI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Specifications

Product Type Drum
Closure Included Yes
Closure Type Screw-on Lid
Capacity (English) 20 gal.
Capacity (Metric) 75.8 L
Color Beige
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Height (English) 20 in.
Height (Metric) 50.8 cm
Weight (English) 13 lb.
Show More Show Less