missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum, 40 gal.
GREENER_CHOICE

Numéro de catalogue. 19338462
Change view
Click to view available options
Color:
Black
Blue
Yellow
Closure Type:
Metal Lever-lock
Plastic Lever-lock
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Color Closure Type
19338462 Yellow Metal Lever-lock
19338461 Black Plastic Lever-lock
19338460 Blue Plastic Lever-lock
19338459 Yellow Plastic Lever-lock
19338464 Black Metal Lever-lock
19338463 Blue Metal Lever-lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. 19338462 Fournisseur Eagle™ Code fournisseur 1651M
Il en reste null

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Spécifications

Product Type Drum
Closure Included Yes
Closure Type Metal Lever-lock
Capacity (English) 40 gal.
Capacity (Metric) 151.6 L
Color Yellow
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Height (English) 29.8 in.
Height (Metric) 98 cm
Weight (English) 22 lb.
Afficher plus de résultats Afficher moins de résultats