missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Lab Pack Open Head Poly Drum, 30 gal.

Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Spécifications
Spécifications
| Product Type | Drum |
| Closure Included | Yes |
| Closure Type | Metal Lever-lock |
| Capacity (English) | 30 gal. |
| Capacity (Metric) | 113.7 L |
| Color | Black |
| For Use With (Application) | Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration |
| Material | HDPE |
| Head Style | Open |
| Height (English) | 26.38 in. |
| Afficher plus de résultats |