missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum with 2 in. Bung Holes, 55 gal.
GREENER_CHOICE

Catalog No. 19338480
Change view
Click to view available options
Color:
Black
Blue
Yelllow
Yellow
Closure Type:
Metal Bolt Ring
Metal Lever-lock
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
19338480 Black Metal Bolt Ring
19338479 Blue Metal Bolt Ring
19338478 Yellow Metal Bolt Ring
19338477 Black Metal Lever-lock
19338476 Blue Metal Lever-lock
19338475 Yelllow Metal Lever-lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19338480 Supplier Eagle™ Supplier No. 1656MBKBRBG2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two, 2 in. holes and is ideal for safe storage, as well as handling or transportation of hazardous materials.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • Bung lid includes two, 2 in. holes
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Specifications

Product Type Drum
Closure Included Yes
Closure Type Metal Bolt Ring
Capacity (English) 55 gal.
Capacity (Metric) 208.48 L
Color Black
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Height (English) 35.8 in.
Height (Metric) 91 cm
Diameter (English) Opening 2 in. (Bung Holes)
Weight (English) 23.5 lb.
Show More Show Less