missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Xylose 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 150°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 150.13 |
| Percent Purity | ≥97.0% (T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-(-)-Xylose |
Safety and Handling
| EINECSNumber | (9)-0318 |
| TSCA | Yes |