missing translation for 'onlineSavingsMsg'
Learn More

L-Valine Methyl Ester Hydrochloride 98.5+%, TCI America™

Catalog No. V00565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00565G 5 g
V005625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00565G Supplier TCI America Supplier No. V00565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6306-52-1
Molecular Formula C6H13ClNO2
Molecular Weight (g/mol) 166.63
MDL Number MFCD00012497
InChI Key RFMIEHOYULVMNZ-UHFFFAOYNA-N
Synonym l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride
PubChem CID 111190
IUPAC Name methyl 2-amino-3-methylbutanoate hydrochloridyl
SMILES [Cl].COC(=O)C(N)C(C)C

Specifications

Color White
Quantity 5 g
Formula Weight 167.63
Percent Purity ≥98.5% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Valine Methyl Ester Hydrochloride