missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine Benzyl Ester Hydrochloride 98.0+%, TCI America™
Supplier: TCI America V00785G
Specifications
| 138°C | |
| 2462-34-2 | |
| C12H18ClNO2 | |
| l-valine benzyl ester hydrochloride, val-obzl hcl, h-val-obzl.hcl, s-benzyl 2-amino-3-methylbutanoate hydrochloride, valine benzyl ester hydrochloride, benzyl 2s-2-amino-3-methylbutanoate hydrochloride, val-obzl.hcl, val-obzl. hcl, l-valine benzyl ester hydrochloride | |
| Cl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 11601206 | |
| ≥98.0% (HPLC,N) |
| L-Valine Benzyl Ester Hydrochloride | |
| White | |
| MFCD22422648 | |
| ZIUNABFUHGBCMF-MERQFXBCSA-N | |
| benzyl (2S)-2-amino-3-methylbutanoate hydrochloride | |
| 243.73 | |
| 243.73 | |
| Crystalline Powder |
Chemical Identifiers
| 2462-34-2 | |
| 243.73 | |
| ZIUNABFUHGBCMF-MERQFXBCSA-N | |
| 11601206 | |
| Cl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1 |
| C12H18ClNO2 | |
| MFCD22422648 | |
| l-valine benzyl ester hydrochloride, val-obzl hcl, h-val-obzl.hcl, s-benzyl 2-amino-3-methylbutanoate hydrochloride, valine benzyl ester hydrochloride, benzyl 2s-2-amino-3-methylbutanoate hydrochloride, val-obzl.hcl, val-obzl. hcl, l-valine benzyl ester hydrochloride | |
| benzyl (2S)-2-amino-3-methylbutanoate hydrochloride |
Safety and Handling
TSCA : No