missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Tyrosine 98.5+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Formula Weight | 181.19 |
| Percent Purity | ≥98.5% (T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-(-)-Tyrosine |
Safety and Handling
| EINECSNumber | (9)-1596 |
| RTECSNumber | YP2275600 |
| TSCA | Yes |