missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111014842
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014842 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014842 Supplier Merck Emd Millipore Supplier No. PHR10971G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 60-18-4
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
MDL Number MFCD00002606
Melting Point >300°C (decomposition)
Synonym (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid; 3-(4-Hydroxyphenyl)-L-alanine
Quantity 1 g
Formula Weight 181.19
Grade Certified Reference Material