missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine, MP Biomedicals™
Specifications
Specifications
| Color | White |
| Assay Percent Range | 0.97 |
| Density | 1.4g/mL |
| Quantity | 1 kg |
| Formula Weight | 181.19 |
| Percent Purity | 99.5% |
| Grade | Research |
| Physical Form | Powder |
| Chemical Name or Material | L-Tyrosine |
Safety and Handling
| EINECSNumber | 200-460-4 |
| Recommended Storage | Store at Room Temperature (15 to 30°C). |