missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111014841
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014841 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014841 Supplier Merck Emd Millipore Supplier No. 91515100MG

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Amino acid precursor of dopamine and other catecholamines

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 60-18-4
Product Line TraceCERT
Physical Form Neat
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
MDL Number MFCD00002606
Melting Point >300°C (decomposition)
Synonym (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid; 3-(4-Hydroxyphenyl)-L-alanine
Quantity 100 mg
Formula Weight 181.19
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC