missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine, Cell Culture Reagent

Catalog No. AAJ63511A1
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63511A1 1 kg
AAJ6351109 10 g
AAJ6351122 100 g
AAJ6351136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ63511A1 Supplier Thermo Scientific Chemicals Supplier No. J63511A1

CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l
PubChem CID 6057
ChEBI CHEBI:17895
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point >300°C
Color Brown to White
Beilstein 392441
Merck Index 14,9839
IUPAC Name 2-amino-3-(4-hydroxyphenyl)propanoic acid
Quantity 1 kg
Formula Weight 181.19
Optical Rotation −11° (c=4 in 1N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Tyrosine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-460-4
RTECSNumber YP2275600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only