missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tryptophan Methyl Ester Hydrochloride 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 220°C |
| Quantity | 5 g |
| Formula Weight | 254.71 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Tryptophan Methyl Ester Hydrochloride |
Sécurité et manipulation
| TSCA | No |