Learn More
Thermo Scientific Chemicals L(-)-Tryptophan, 99%
L(-)-Tryptophan, CAS # 73-22-3, is an essential amino acid necessary for the biosynthesis of proteins. | CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol
Supplier: Thermo Scientific Chemicals 140590250
| Quantity | 25 g |
|---|---|
| Packaging | Plastic Bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- L(-)-Tryptophan is a nonpolar aromatic compound and essential amino acid required for protein synthesis
- It is a biochemical precursor of niacin, indole alkaloids, and serotonin
Applications
- L-Tryptophan acts as a natural dietary supplement and has laboratory applications in the research of antidepressants, anxiolytics, and sleep aids
- It is a precursor of metabolites that are important in the laboratory study of mammalian physiology, including gastrointestinal functions, immunity, metabolism, and the nervous system
Chemical Identifiers
| C11H12N2O2 | |
| MFCD00064340 | |
| l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin | |
| CHEBI:16828 | |
| N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O |
Specifications
| 280.0°C to 285.0°C | |
| 99% | |
| White to White-Yellow | |
| 0.5% max. (1 g, 105°C) | |
| Plastic Bottle | |
| MFCD00064340 | |
| 22, 546 | |
| l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin | |
| QIVBCDIJIAJPQS-VIFPVBQESA-N | |
| − 31.10 (20.00°C c=1,H20) | |
| 204.23 | |
| 6305 | |
| 204.23 | |
| Crystalline Powder |
| L(-)-Tryptophan | |
| 73-22-3 | |
| Authentic | |
| 99% | |
| C11H12N2O2 | |
| − 31.10 | |
| 15, 9977 | |
| Solubility in water: 11.4g/L (25°C). Other solubilities: soluble in alkali hydroxides and hot alcohol, insoluble in chloroform | |
| N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O | |
| (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid | |
| 25 g | |
| CHEBI:16828 | |
| 99% |
Safety and Handling
EINECSNumber : 200-795-6
RTECSNumber : YN6130000
TSCA : TSCA