missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015352
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015352 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015352 Supplier Merck Emd Millipore Supplier No. PHR1242500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-Threonine is an essential amino acid. It is involved in the synthesis of mucin, a protein supporting intestinal function and integrity. L-threonine has a role in the immune system, in O-linked glycosylation, protein phosphorylation and glycine synthesis.

Specifications

CAS 72-19-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula CH3 CH(OH)CH(NH2)CO2 H
MDL Number MFCD00064270
Melting Point 256°C
Synonym (2 S,3 R)-2-Amino-3-hydroxybutyric acid
Type Certified Reference Material
Quantity 500 mg
Formula Weight 119.12
Grade Certified Reference Material
Show More Show Less