missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Threonine, 98%

Numéro de catalogue. p-3734537
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC138930050 5 g Glass Bottle
AC138930250 25 g Glass Bottle
AC138931000 100 g Glass Bottle
AC138935000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. AC138930050 Fournisseur Thermo Scientific Chemicals Code fournisseur 138930050
Il en reste null

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
SMILES CC(O)C(N)C(O)=O

Spécifications

Melting Point 255.0°C
Color Yellow
Chloride 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Loss on Drying 0.5 % max. (1 g, 105°C, 3 hrs)
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH2)CO2H
Specific Rotation − 28.30
Sulfate 300ppm max.
Merck Index 15, 9534
Sulfated Ash 0.1 % max.
Solubility Information Solubility in water: 90g/L (20°C). Other solubilities: insoluble in most common organic solvents
IUPAC Name 2-amino-3-hydroxybutanoic acid
Specific Rotation Condition −28.30 (20.00°C c=6,H2O)
Quantity 5 g
Formula Weight 119.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Threonine
Afficher plus de résultats Afficher moins de résultats

RUO – Research Use Only