missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine methyl ester hydrochloride, 97%

Catalog No. AAH5956303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5956303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5956303 Supplier Thermo Scientific Chemicals Supplier No. H5956303
Only null left

CAS: 63038-27-7 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63038-27-7
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00040604
InChI Key HRTQWUHFSXVRPY-XZNNNFJINA-N
Synonym l-tert-leucine methyl ester hydrochloride, h-tle-ome.hcl, s-methyl 2-amino-3,3-dimethylbutanoate hydrochloride, methyl 2s-2-amino-3,3-dimethylbutanoate hydrochloride, s-2-amino-3,3-dimethylbutyric acid methyl ester hydrochloride, methyl 3-methyl-l-valinate hydrochloride, pubchem11076, methyl l-tert-leucinate hydrochloride, l-tert-leucine methyl ester hcl, l-tert-leucine methyl-ester hydrochloride
PubChem CID 16217630
IUPAC Name methyl (2S)-2-amino-3,3-dimethylbutanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)C(C)(C)C

Specifications

Melting Point 183°C
Beilstein 4879785
Quantity 1 g
Formula Weight 181.66
Optical Rotation +18° (c=1 in Methanol)
Percent Purity 97%
Chemical Name or Material L-tert-Leucine methyl ester hydrochloride
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only