missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Sorbose 98.0+%, TCI America™

Catalog No. S006625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S006625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S006625G Supplier TCI America Supplier No. S006625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-79-6
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00151097
InChI Key BJHIKXHVCXFQLS-OTWZMJIISA-N
Synonym l---sorbose, 3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, sorbinose, keto-l-sorbose, esorben, sorbin, l--sorbose, hsdb 780, l-sorbinose, l-xylo-2-hexulose
PubChem CID 6904
ChEBI CHEBI:13172
IUPAC Name (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 165°C
Color White
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Sorbose
EINECSNumber (8)-0521
RTECSNumber WG3195025
TSCA Yes