missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine, Cell Culture Reagent

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol

$69.12 - $1,488.52

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064224
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6218709
View Documents Promotion Details
Thermo Scientific Chemicals
J6218709
10 g
Each for $69.12
Only null left
 
AAJ6218722
View Documents Promotion Details
Thermo Scientific Chemicals
J6218722
100 g
Each for $214.14
Only null left
 
AAJ6218736
View Documents Promotion Details
Thermo Scientific Chemicals
J6218736
500 g
Each for $748.62
Only null left
 
AAJ62187A1
View Documents Promotion Details
Thermo Scientific Chemicals
J62187A1
1 kg
Each for $1,488.52
Only null left
 
Description

Description

It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

Solubility
Soluble in water.

Notes
Store in cool, dry place. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

56-45-1
105.093
MTCFGRXMJLQNBG-REOHCLBHSA-N
5951
(2S)-2-amino-3-hydroxypropanoic acid
C3H7NO3
MFCD00064224
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
CHEBI:17115
C(C(C(=O)O)N)O

Specifications

222°C
White
MFCD00064224
14,8460
Soluble in water.
C(C(C(=O)O)N)O
105.093
CHEBI:17115
Cell Culture Reagent
L-Serine
56-45-1
C3H7NO3
1721404
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
MTCFGRXMJLQNBG-REOHCLBHSA-N
(2S)-2-amino-3-hydroxypropanoic acid
5951
105.09
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-274-3

RTECSNumber : VT8100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only